missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ dGTP BigDye™ Terminator v1.0 Ready Reaction Cycle Sequencing Kit

Product Code. 15839746 Shop All Applied Biosystems Products
Click to view available options
Quantity:
100 Reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15839746

Brand: Applied Biosystems™ 4307175

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Contains AmpliTaq™ DNA Polymerase, FS, BigDye terminators v1.0, pGEM™ - 3Zf(+) Control Template, -21 M13 Control Primer (forward), and Premixed Ready Reaction.

dGTP BigDye™ terminator cycle sequencing kit with the AmpliTaq™ DNA polymerase, FS. This ready reaction kit uses the same dye set as the BigDye™ v1.0 kits.

  • Ensure reliable results with difficult regions and finishing applications
  • Sequence templates with challenging sequence contexts
  • Sequence problematic templates that standard chemistries cannot
  • Optimized for GT-Rich templates: Obtain higher success rate even with your difficult GT-rich templates
  • By using the same chemistries as the standard BigDye™ terminator kits (only replacing dITP with dGTP) the dGTP BigDye™ terminator kit provides proven performance
  • Optimized for GT-rich and other difficult-to-sequence templates
  • Easily Integrate into your workflow: dGTP BigDye™ terminator kits require no new software or instrument recalibration, so you can easily integrate it into your current workflow with minimal changes to your protocols
  • The dGTP BigDye™ terminator kit replaces dITP with dGTP and it enables you to extend through those difficult-to-sequence regions with confidence, avoiding early signal loss in these samples

Order Info

100 rxns

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The kit contains the AmpliTaq™ DNA Polymerase, FS, BigDye
terminators v1.0, pGEM™ - 3Zf(+) Control Template,
-21 M13 Control Primer (forward), and Premixed Ready Reaction.
Store kit at -15 to -25°C.
Format 12-well Strips, 8-well Strip, 96- or 384-well Plate
For Use With (Application) GT-Rich or Difficult Template Sequencing
No. of Reactions 100 reactions
Product Line BigDye Terminator, BigDye
Product Type Sequencing Kit
Quantity 100 Reactions
Technique Fluorescent Dye Terminator Sequencing
Template Compatibility BAC DNA, Genomic DNA (Bacterial), Plasmid DNA (≤15Kb), PCR Amplicons, Single Stranded DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.