missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DGKQ Polyclonal Antibody
GREENER_CHOICE

Product Code. 15507621
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15507621 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15507621 Supplier Invitrogen™ Supplier No. PA536313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 101 kDa. Purity is >95% by SDS-PAGE.

Diacylglycerol (DAG) is a fundamental lipid second messenger that is produced in the nucleus. The accumulation of DAG in the nucleus is important for the regulation of cell growth and differentiation. Diacylglycerol kinases (DGKs) convert DAG to phosphatidic acid, thereby terminating diacylglycerol signaling, which results in the reduction of protein kinase C activity and cell cycle progression of T lymphocytes. Diacylglycerol kinases are divided into five subtypes, Type I-Type V. DGK-theta is a Type V DGK, and localizes mainly to the nucleus of various cell lines, such as MDA-MB-453, MCF-7, PC12 and HeLa. Nuclear DGK-theta co-localizes with phosphatidylinositol 4, 5-bisphosphate (PIP(2)). DGK-theta is the isoform responsive to alpha-Thrombin stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGKQ
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene DGKQ
Gene Accession No. P52824
Gene Alias 110kDa; DAG kinase theta; DAGK; Dagk4; DAGK7; Dgk theta; Dgkd; DGKQ; DGK-theta; Diacylglycerol kinase theta; diacylglycerol kinase, delta (110 kDa); diacylglycerol kinase, theta; diacylglycerol kinase, theta 110kDa; diglyceride kinase theta; testis tissue sperm-binding protein Li 38a
Gene Symbols DGKQ
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 700-750 of human DGK-theta.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1609
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.