missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGKK Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | DGKK |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16819574
|
Proteintech
20090-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16809574
|
Proteintech
20090-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to DGKK.
DGKK phosphorylates diacylglycerol (DAG) to generate phosphatidic acid (PA).Specifications
| DGKK | |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q5KSL6 | |
| 139189 | |
| Peptide | |
| Primary | |
| -20°C | |
| DGKK |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 142 kDa diacylglycerol kinase, DAG kinase kappa, DGK kappa, DGKK, Diacylglycerol kinase kappa, diacylglycerol kinase, kappa, Diglyceride kinase kappa | |
| DGKK | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title