missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGKH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13873-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the diacylglycerol kinase (DGK) enzyme family. Members of this family are involved in regulating intracellular concentrations of diacylglycerol and phosphatidic acid. Variation in this gene has been associated with bipolar disorder. Alternatively spliced transcript variants have been identified.Specifications
| DGKH | |
| Polyclonal | |
| Unconjugated | |
| DGKH | |
| DAG kinase eta, DGK eta, DGKeta, DGKH, Diacylglycerol kinase eta, diacylglycerol kinase, eta, Diglyceride kinase eta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 160851, 361076, 380921 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86XP1 | |
| Dgkh | |
| DGKH Fusion Protein Ag4926 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction