missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DGKH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558829
This item is not returnable.
View return policy
Description
Immunogen sequence: VRQVIEEAGK VMDDPTVHPC EPANQSSDYD STETDESKEE AKDDGAKESI TVKTAPRSPD ARASYGHSQT DSVPGPAVAA SKENLPVLN Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 69%.
This gene encodes a member of the diacylglycerol kinase (DGK) enzyme family. Members of this family are involved in regulating intracellular concentrations of diacylglycerol and phosphatidic acid. Variation in this gene has been associated with bipolar disorder. Alternatively spliced transcript variants have been identified.
Specifications
| DGKH | |
| Polyclonal | |
| Unconjugated | |
| Dgkh | |
| 5930402B05Rik; D130015C16; DAG kinase eta; DGK; DGKeta; DGK-eta; DGKH; Diacylglycerol kinase eta; diacylglycerol kinase, eta; diglyceride kinase eta; DKFZp761I1510; EC 2.7.1.107; RGD1561955; si:ch211-93a19.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 160851 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86XP1 | |
| Dgkh | |
| Recombinant protein corresponding to Human DGKH. Recombinant protein control fragment (Product #RP-97925). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction