missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGK-zeta Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97260PE
This item is not returnable.
View return policy
Description
DGK-zeta Polyclonal antibody specifically detects DGK-zeta in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DGK-zeta | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| DAGK5, DAGK6DAG kinase zeta, DGK-ZETA, diacylglycerol kinase zeta, diacylglycerol kinase, zeta, diacylglycerol kinase, zeta 104kDa, Diglyceride kinase zeta, EC 2.7.1.107, hDGKzeta | |
| Produced in rabbits immunized with E. coli-derived Human DGK-zeta fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 8525 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction