missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGK-theta Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92780-0.1ml
This item is not returnable.
View return policy
Description
DGK-theta Polyclonal antibody specifically detects DGK-theta in Human, Mouse samples. It is validated for Western Blot
Specifications
| DGK-theta | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000 | |
| DAG kinase theta, DAGK, DAGK4EC 2.7.1.107, DAGK7, DGK-theta, diacylglycerol kinase theta, diacylglycerol kinase, theta (110kD), diacylglycerol kinase, theta 110kDa, Diglyceride kinase theta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 683-942 of human DGKQ (NP_001338.2). GRVLRWGAGYSGEDPFSVLLSVDEADAVLMDRWTILLDAHEAGSAENDTADAEPPKIVQMSNYCGIGIDAELSLDFHQAREEEPGKFTSRLHNKGVYVRVGLQKISHSRSLHKQIRLQVERQEVELPSIEGLIFINIPSWGSGADLWGSDSDTRFEKPRMDDGLLEVVGVTGVVHMGQVQGGLRSGIRIAQGSYFRVTLLKATPVQVDGEPWVQAPGHMIISAAGPKVHMLRKAKQKPRRAGTTRDARADAAPAPESDPR | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1609 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction