missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGK-gamma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33055
This item is not returnable.
View return policy
Description
DGK-gamma Polyclonal specifically detects DGK-gamma in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DGK-gamma | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| DGKG | |
| Recombinant protein encompassing a sequence within the center region of human DGK-gamma. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Expected to cross react based on sequence identity: Mouse (95%), Rat (96%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.92mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| DAG kinase gamma, DAGK3MGC104993, DGK-GAMMA, diacylglycerol kinase gamma, diacylglycerol kinase, gamma (90kD), diacylglycerol kinase, gamma 90kDa, diacylglyerol kinase gamma, Diglyceride kinase gamma, EC 2.7.1.107, MGC133330 | |
| Rabbit | |
| 89 kDa | |
| 0.1 mL | |
| Lipid and Metabolism, Vision | |
| 1608 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction