missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGK-epsilon Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85315
This item is not returnable.
View return policy
Description
DGK-epsilon Polyclonal specifically detects DGK-epsilon in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DGK-epsilon | |
| Polyclonal | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| DAG kinase epsilon, DAGK5, DAGK6, DGK, DGK-epsilon, diacylglycerol kinase epsilon, diacylglycerol kinase, epsilon (64kD), diacylglycerol kinase, epsilon 64kDa, Diglyceride kinase epsilon, EC 2.7.1.107 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DGKE | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RDTDLFSQPTYCCVCAQHILQGAFCDCCGLRVDEGCLRKADKRFQCKEIMLKNDTKVLDAMPHHWIRGNVPLCSYCMVCKQQCGCQPKLCDYRCIWCQKTVHDECMKNSLKNEKCDFGEFKNLIIPPSYL | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 8526 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction