missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGK-delta Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92207-0.1ml
This item is not returnable.
View return policy
Description
DGK-delta Polyclonal antibody specifically detects DGK-delta in Human samples. It is validated for Western Blot
Specifications
| DGK-delta | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 130 kDa diacylglycerol kinase, DAG kinase delta, dgkd-2, DGKdelta, DGK-delta, diacylglycerol kinase delta, diacylglycerol kinase, delta (130kD), diacylglycerol kinase, delta 130kDa, Diglyceride kinase delta, FLJ26930, KIAA0145EC 2.7.1.107 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 960-1140 of human DGKD (NP_690618.2). LPRPPSCSLHPEMLSEEEATQMDQFGQAAGVLIHSIREIAQSHRDMEQELAHAVNASSKSMDRVYGKPRTTEGLNCSFVLEMVNNFRALRSETELLLSGKMALQLDPPQKEQLGSALAEMDRQLRRLADTPWLCQSAEPGDEESVMLDLAKRSRSGKFRLVTKFKKEKNNKNKEAHSSLGA | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 8527 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction