missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGK-alpha Antibody (OTI4A11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70547CL1
This item is not returnable.
View return policy
Description
DGK-alpha Monoclonal antibody specifically detects DGK-alpha in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),CyTOF
Specifications
| DGK-alpha | |
| Monoclonal | |
| CoraFluor 1 | |
| DAG kinase alpha, DAGK, DAGK1diacylglycerol kinase, alpha (80kD), DGK-alphaEC 2.7.1.107, diacylglycerol kinase alpha, diacylglycerol kinase, alpha 80kDa, Diglyceride kinase alpha, MGC12821,80 kDa diacylglycerol kinase, MGC42356 | |
| Full length human recombinant protein of human DGKA(NP_001336) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1606 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), CyTOF | |
| OTI4A11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction