missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DGCR8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540122
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, and HepG2 cells. Predicted reactivity: Mouse (82%), Rat (80%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a subunit of the microprocessor complex which mediates the biogenesis of microRNAs from the primary microRNA transcript. The encoded protein is a double-stranded RNA binding protein that functions as the non-catalytic subunit of the microprocessor complex. This protein is required for binding the double-stranded RNA substrate and facilitates cleavage of the RNA by the ribonuclease III protein, Drosha. Alternate splicing results in multiple transcript variants.
Specifications
| DGCR8 | |
| Polyclonal | |
| Unconjugated | |
| DGCR8 | |
| C22orf12; D16H22S1742E; D16H22S788E; D16Wis2; DGCR8; DGCR8 microprocessor complex subunit; DGCR8, microprocessor complex subunit; DGCRK6; DiGeorge syndrome critical region 8; diGeorge syndrome critical region 8 homolog; DiGeorge syndrome critical region gene 8; Gy1; LP4941; Microprocessor complex subunit DGCR8; mir-1306; N41; pasha; sb:eu827; sb:eu832; si:ch211-106a19.4; Vo59c07; wu:fc23f08; wu:fc38f06 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54487 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.15 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q8WYQ5 | |
| DGCR8 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human DGCR8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Amphibian, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction