missing translation for 'onlineSavingsMsg'
Learn More

DGCR8 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18446422 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18446422 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18446422 Supplier Novus Biologicals Supplier No. NBP130116AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DGCR8 Polyclonal antibody specifically detects DGCR8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGCR8
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C22orf12, DGCRK6chromosome 22 open reading frame 12, DiGeorge syndrome critical region 8, DiGeorge syndrome critical region gene 8, Gy1, microprocessor complex subunit DGCR8, pasha
Host Species Rabbit
Immunogen Antigen maps to a region between residue 400 and 450 of human DiGeorge syndrome critical region gene 8 using the numbering given in entry NP_073557.3 (GeneID 54487).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 54487
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.