missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGCR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13705-1-AP-150UL
This item is not returnable.
View return policy
Description
Deletions of the 22q11.2 have been associated with a wide range of developmental defects (notably DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome and isolated conotruncal cardiac defects) classified under the acronym CATCH 22. The DGCR2 gene encodes a novel putative adhesion receptor protein, which could play a role in neural crest cells migration, a process which has been proposed to be altered in DiGeorge syndrome.Specifications
| DGCR2 | |
| Polyclonal | |
| Unconjugated | |
| DGCR2 | |
| DGCR2, DGS C, IDD, KIAA0163, LAN, SEZ 12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13356, 360742, 9993 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P98153, P98154 | |
| Dgcr2 | |
| DGCR2 Fusion Protein Ag4629 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction