missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGCR14 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | DGCR14 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DGCR14 Polyclonal antibody specifically detects DGCR14 in Human, Mouse samples. It is validated for Western BlotSpecifications
| DGCR14 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 8220 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DGCR13, DGS-HDGS-I, DGSIDGSH, DiGeorge syndrome critical region 13, DiGeorge syndrome critical region 14, DiGeorge syndrome critical region gene 13, DiGeorge syndrome critical region gene 14, DiGeorge syndrome gene H, DiGeorge syndrome gene I, DiGeorge syndrome protein H, ES2DiGeorge syndrome critical region gene DGSI, Es2el, Protein DGCR13, protein DGCR14, Protein ES2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human DGCR14 (NP_073210.1). METPGASASSLLLPAASRPPRKREAGEAGAATSKQRVLDEEEYIEGLQTVIQRDFFPDVEKLQAQKEYLEAEENGDLERMRQIAIKFGSALGKMSREPPPPYVTPATFETPEVHAGTGVVGNKPRPRGRGLEDGEAGEEEEKEPLPSLDVFLSRYTSEDNASFQEIMEVAKERSRARHAWLYQAEEEFEKRQKDNLELPSAEHQAIESSQASVETWKYKAKNSLMYYPEGVPDEEQLFKKPRQVVHKNTRFLRDPFSQALSRCQLQQAAALNAQHKQGKVGPDGKELIPQESPRVGGFGF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title