missing translation for 'onlineSavingsMsg'
Learn More

DGAT2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18421833
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421833 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421833 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP171701MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DGAT2 Polyclonal antibody specifically detects DGAT2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGAT2
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias diacylglycerol O-acyltransferase 2, diacylglycerol O-acyltransferase homolog 2, diacylglycerol O-acyltransferase homolog 2 (mouse), diacylglycerol O-acyltransferase-like protein 2, Diglyceride acyltransferase 2, DKFZp686A15125, EC 2.3.1, EC 2.3.1.20, GS1999FULL
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human DGAT2 protein (within residues 200-300). [Swiss-Prot Q96PD7]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 84649
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.