missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DGAT2 Polyclonal antibody specifically detects DGAT2 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
Specifications
Specifications
| Antigen | DGAT2 |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | diacylglycerol O-acyltransferase 2, diacylglycerol O-acyltransferase homolog 2, diacylglycerol O-acyltransferase homolog 2 (mouse), diacylglycerol O-acyltransferase-like protein 2, Diglyceride acyltransferase 2, DKFZp686A15125, EC 2.3.1, EC 2.3.1.20, GS1999FULL |
| Host Species | Sheep |
| Immunogen | E. coli-derived recombinant human DGAT2, Arg268-Phe377, Accession # Q96PD7 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?