missing translation for 'onlineSavingsMsg'
Learn More

DGAT2 Antibody [DyLight 650], Novus Biologicals™

Product Code. 30736064 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736064 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736064 Supplier Novus Biologicals Supplier No. AF7256W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

DGAT2 Polyclonal antibody specifically detects DGAT2 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGAT2
Applications Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias diacylglycerol O-acyltransferase 2, diacylglycerol O-acyltransferase homolog 2, diacylglycerol O-acyltransferase homolog 2 (mouse), diacylglycerol O-acyltransferase-like protein 2, Diglyceride acyltransferase 2, DKFZp686A15125, EC 2.3.1, EC 2.3.1.20, GS1999FULL
Host Species Sheep
Immunogen E. coli-derived recombinant human DGAT2, Arg268-Phe377, Accession # Q96PD7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 84649
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.