missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGAT1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA5142774
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes.Specifications
DGAT1 | |
Polyclonal | |
Unconjugated | |
DGAT1 | |
ACAT related gene product 1; ACAT-related gene product 1; Acyl coenzyme A; acyl coenzyme A:cholesterol acyltransferase related gene 1; acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; acyl-coenzyme A:diacylglycerol acyltransferase 1; AGRP1; ARAT; ARGP1; C75990; D15Ertd23e; Dgat; DGAT1; dgat1a; DGAT1B; diacylglycerol acyltransferase; diacylglycerol acyltransferase 1; diacylglycerol O-acyltransferase 1; diacylglycerol O-acyltransferase 1a; diacylglycerol O-acyltransferase homolog 1; diacylglycerol O-acyltransferase homolog 1a; DIAR7; Diglyceride acyltransferase; EC 2.3.1.20; hypothetical protein LOC325875; putative diacylglycerol acyltransferase 1; retinol O-fatty-acyltransferase; wu:fd36g11; zgc:77691 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
13350 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9Z2A7 | |
DGAT1 | |
Peptide with sequence RCHRLQDSLFSSD. | |
100 μg | |
Primary | |
Mouse | |
Liquid |