missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DGAT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11326613
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326613 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11326613 Supplier Invitrogen™ Supplier No. PA518634

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGAT1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DGAT1
Gene Accession No. O75907, Q9Z2A7
Gene Alias ACAT related gene product 1; ACAT-related gene product 1; Acyl coenzyme A; acyl coenzyme A:cholesterol acyltransferase related gene 1; acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; acyl-coenzyme A:diacylglycerol acyltransferase 1; AGRP1; ARAT; ARGP1; C75990; D15Ertd23e; Dgat; DGAT1; dgat1a; DGAT1B; diacylglycerol acyltransferase; diacylglycerol acyltransferase 1; diacylglycerol O-acyltransferase 1; diacylglycerol O-acyltransferase 1a; diacylglycerol O-acyltransferase homolog 1; diacylglycerol O-acyltransferase homolog 1a; DIAR7; Diglyceride acyltransferase; EC 2.3.1.20; hypothetical protein LOC325875; putative diacylglycerol acyltransferase 1; retinol O-fatty-acyltransferase; wu:fd36g11; zgc:77691
Gene Symbols DGAT1
Host Species Goat
Immunogen Synthetic peptide sequence (QNSMKPFKDMDYS) corresponding to the internal amino acids of DGAT1 (aa 305-317).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13350, 8694
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.