missing translation for 'onlineSavingsMsg'
Learn More

DGAT1-IN-1, MedChemExpress

Product Code. 16497877
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497877 5 mg 5mg
16455158 10 mg 10mg
16425866 100 mg 100mg
16401387 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497877 Supplier Medchem Express Supplier No. HY12425
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DGAT1-IN-1 is a potent DGAT1 inhibitor with IC50 of < 10 nM(cell lysate from Hep3B cells overexpressing human DGAT1).

DGAT1-IN-1 is a potent DGAT1 inhibitor with IC50 of < 10 nM(cell lysate from Hep3B cells overexpressing human DGAT1).

Specifications

Chemical Name or Material DGAT1-IN-1
CAS 1449779-49-0
Color White
Purity Grade Notes Research
Molecular Formula C30H28F3N3O4
Quantity 5 mg
Solubility Information DMSO : 20 mg/mL (36.26 mM; Need ultrasonic)
SMILES OC(C[C@H]1CC[C@H](C2=CC=C(C3=CN4C=C(C(NCC5=CC=C(OC(F)(F)F)C=C5)=O)N=C4C=C3)C=C2)CC1)=O
Molecular Weight (g/mol) 551.56
Formula Weight 551.56
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.