missing translation for 'onlineSavingsMsg'
Learn More

DG051, MedChemExpress

Product Code. 16456887
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456887 5 mg 5mg
16491099 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16456887 Supplier Medchem Express Supplier No. HY10825
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DG051 is a potent leukotriene A4 hydrolase inhibitor of leukotriene B4 biosynthesis in the enzyme assay with an IC50=47 nM.

DG051 is a potent leukotriene A4 hydrolase inhibitor of leukotriene B4 biosynthesis in the enzyme assay with an IC50=47 nM.

Specifications

Chemical Name or Material DG051
CAS 929915-58-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H25Cl2NO4
Quantity 5 mg
Solubility Information DMSO : ≥ 325 mg/mL (762.32 mM)
SMILES Cl[H].O=C(O)CCCN1CCC[C@H]1COC(C=C2)=CC=C2OC3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 426.33
Formula Weight 426.33
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.