missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DFNA5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13075-1-AP-150UL
This item is not returnable.
View return policy
Description
DFNA5 (deafness, autosomal domit 5), also known as ICERE-1, is a 496 amino acid protein that is expressed in cochlea tissue, as well as in placenta, brain, heart, liver, lung and pancreas as two alternatively spliced isoforms, designated short and long. Defects in the gene encoding DFNA5 are the cause of non-syndromic sensorineural deafness autosomal domit type 5 (DFNA5), a form of sensorineural hearing loss that results from damage to one of various structures that receive sound information in the brain. The gene encoding DFNA5 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.Specifications
| DFNA5 | |
| Polyclonal | |
| Unconjugated | |
| DFNA5 | |
| deafness, autosomal domit 5, DFNA5, GSDME, ICERE 1, ICERE1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1687, 54722 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60443, Q9Z2D3 | |
| DFNA5 | |
| DFNA5 Fusion Protein Ag3746 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction