missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DFNA5 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | DFNA5 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC019689 |
| Molecular Weight (g/mol) | Theoretical M.W. 32.56kDa |
| Plasmid | pCMV-DFNA5 full-length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-DFNA5 antibody (Mfr. No. H00001687-B01) by Western Blot. |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Host Species | Human |
| Show More |
Safety and Handling
Product Identifier
- 100UL DFNA5 293T Cell Transient Overexpression Lys
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contain: Mercaptoethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?