missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DFMO
GREENER_CHOICE

Product Code. 18014722
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18014722

Brand: Tocris Bioscience™ 2761/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ornithine decarboxylase inhibitor

Irreversible inhibitor of ornithine decarboxylase (ODC) that inhibits polyamine biosynthesis. Displays antiapoptotic, antiangiogenic and antiparasitic activity. Induces re-expression of aberrantly silenced tumor suppressor genes when used in combination with oligoamine analogs.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68278-23-9
Molecular Formula C6H13ClF2N2O2

Specifications

Quantity 10 mg
Formula Weight 218.63
Purity >98%
Molecular Weight (g/mol) 218.629
Chemical Name or Material DFMO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.