missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DFFB Polyclonal Antibody

Product Code. 12808112
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808112 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808112 Supplier Invitrogen™ Supplier No. PA519892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~40kDa. A suggested positive control is mouse lung tissue lysate. PA5-19892 can be used with blocking peptide PEP-0020.

Caspase-activated DNase (CAD) cleaves chromosomal DNA during apoptosis. Apoptosis which is a cell death process removing toxic and/or useless cells during mammalian development can be induced by a variety of stimuli, including the death factors of the tumor necrosis factor family, or UV irradiation, anticancer drugs, and survival factor deprivation. The apoptotic process is accompanied by shrinkage and fragmentation of the cells and nuclei and degradation of the chromosomal DNA into nucleosomal units. CAD, which can be activated by caspases, exists as a complex with its inhibitor, inhibitor of CAD (ICAD). ICAD helps correct folding of CAD and remains complexed with CAD to inhibit the DNase activity of CAD. Caspase, which is activated during the apoptotic process, cleaves ICAD, and the CAD released from ICAD enters the nuclei to cleave the chromosomal DNA. Internal as well as external signals can activate caspases and this is under the control of the balance between pro-and anti-apoptotic proteins of the Bcl-2 family, heat shock proteins, and inhibitors of apoptosis proteins (IAPs). Currently, fourteen members of the caspase family have been identified, of which seven mediate apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DFFB
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DFFB
Gene Accession No. O54788
Gene Alias 2410008J01Rik; 40kDa; 5730477D02Rik; AU018859; CAD; caspase-activated deoxyribonuclease; caspase-activated DNase; caspase-activated nuclease; Cpad; CPAN; DFF2; DFF-2; DFF40; DFF-40; DFFB; Didff; DNA fragmentation factor 40 kD beta polypeptide (caspase-activated DNase); DNA fragmentation factor 40 kDa subunit; DNA fragmentation factor subunit beta; DNA fragmentation factor, 40 kD, beta polypeptide (caspase-activated DNase); DNA fragmentation factor, 40 kD, beta subunit; DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase); DNA fragmentation factor, beta polypeptide (caspase-activated DNase); DNA fragmentation factor, beta subunit; DNase inhibited by DNA fragmentation factor; RP13-531C17.1; RP23-254N4.2
Gene Symbols DFFB
Host Species Rabbit
Immunogen A 16 amino acid peptide near the carboxy terminus of murine CAD.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13368
Target Species Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.