missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DFF40/CAD Antibody (36A349), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-13592IR
This item is not returnable.
View return policy
Description
DFF40/CAD Monoclonal specifically detects DFF40/CAD in Human, Mouse samples. It is validated for Western Blot.
Specifications
| DFF40/CAD | |
| Monoclonal | |
| DyLight 755 | |
| CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.- | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In HeLa, a 40 kDa band is observed. | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| 36A349 | |
| Western Blot | |
| DFFB | |
| Anti-DFF40 monoclonal antibody was raised against a peptide corresponding to amino acids 205 to 222 of human DFF40. | |
| 0.1 mL | |
| Apoptosis | |
| 1677 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction