missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DFF40/CAD Antibody (36A349), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-13592AF532
This item is not returnable.
View return policy
Description
DFF40/CAD Monoclonal specifically detects DFF40/CAD in Human, Mouse samples. It is validated for Western Blot.
Specifications
| DFF40/CAD | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DFFB | |
| Anti-DFF40 monoclonal antibody was raised against a peptide corresponding to amino acids 205 to 222 of human DFF40. | |
| Primary | |
| In HeLa, a 40 kDa band is observed. | |
| Store at 4°C in the dark. |
| Western Blot | |
| 36A349 | |
| Western Blot | |
| CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.- | |
| Mouse | |
| 0.1 mL | |
| 1677 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction