missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DFF40/CAD Antibody (36A349), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-13592AF405
This item is not returnable.
View return policy
Description
DFF40/CAD Monoclonal specifically detects DFF40/CAD in Human, Mouse samples. It is validated for Western Blot.
Specifications
| DFF40/CAD | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DFFB | |
| Anti-DFF40 monoclonal antibody was raised against a peptide corresponding to amino acids 205 to 222 of human DFF40. | |
| 0.1 mL | |
| Apoptosis | |
| 1677 | |
| Human | |
| Purified |
| Western Blot | |
| 36A349 | |
| Western Blot | |
| CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.- | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In HeLa, a 40 kDa band is observed. | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction