missing translation for 'onlineSavingsMsg'
Learn More

Dezocine, TRC

Product Code. 30357495
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.50mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357495 1 mg 1mg
30361294 0.5 mg 0.50mg
30362829 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357495 Supplier Toronto Research Chemicals Supplier No. TRCD2998001MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53648-55-8
Molecular Formula C16H23NO
Molecular Weight (g/mol) 245.36
Synonym (5R,11S,13S)-13-Amino-5,6,7,8,9,10,11,12-octahydro-5-methyl-5,11-methanobenzocyclodecen-3-ol, [5R-(5α,11α,13S*)]-13-Amino-5,6,7,8,9,10,11,12-octahydro-5-methyl-5,11-methanobenzocyclodecen-3-ol, (-)-Dezocine, Wy 16225
SMILES C[C@]12CCCCC[C@H]([C@@H]1N)CC3=C2C=C(C=C3)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H23NO/c1-16-8-4-2-3-5-12(15(16)17)9-11-6-7-13(18)10-14(11)16/h6-7,10,12,15,18H,2-5,8-9,17H2,1H3/t12-,15-,16+/m0/s1
Formula Weight 245.178
Chemical Name or Material Dezocine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.