missing translation for 'onlineSavingsMsg'
Learn More

Dextrose, Monohydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

£420.00

Chemical Identifiers

CAS 1496309
MDL Number MFCD00150899
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15547934
View Documents Promotion Details
Avantor J.T.Baker
1910-05
2.5 kg Poly Bottle
£420.00
2.50kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

1496309
MFCD00150899

Specifications

1496309
MFCD00150899
nicotine ditartrate dihydrate, unii-r7m676m8yv, nicotine bitartrate usan, nicotine bitartrate who-dd, nicotine acid tartrate dihydrate, s-3-1-methylpyrrolidin-2-yl pyridine bis 2r,3r-2,3-dihydroxysuccinate dihydrate, 2r,3r-2,3-dihydroxybutanedioic acid; 3-2s-1-methylpyrrolidin-2-yl pyridine; dihydrate, 3-2s-1-methyl-2-pyrrolidinyl pyridine, 2r,3r-2,3-dihydroxybutanedioate 1:2 , dihydrate
O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1
498.44
BAKER ANALYZED™ Reagent
Dextrose, Monohydrate
C18H30N2O14
1L = 1.54kg
LDMPZNTVIGIREC-HYRPJEHINA-N
(2S,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol hydrate
198.17
Powder
Videos
SDS
Documents

Documents

Product Certifications