missing translation for 'onlineSavingsMsg'
Learn More

Dextrorotation nimorazole phosphate ester, MedChemExpress

Product Code. 16497598
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497598 10 mg 10mg
16493617 50 mg 50mg
16430078 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497598 Supplier Medchem Express Supplier No. HY18716

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dextrorotation nimorazole phosphate ester is an anti-anaerobic and anti-parasitic agent.

Dextrorotation nimorazole phosphate ester is an anti-anaerobic and anti-parasitic agent.

Specifications

Chemical Name or Material Dextrorotation nimorazole phosphate ester
Physical Form Solid
CAS 1124347-33-6
Color Red
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C11H19N4O7P
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 32 mg/mL (91.36 mM)
SMILES CC1=NC=C([N+]([O-])=O)N1C[C@H](OP(O)(O)=O)CN2CCOCC2
Molecular Weight (g/mol) 350.26
Formula Weight 350.26
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.