missing translation for 'onlineSavingsMsg'
Learn More

Dextrin, TRC

Product Code. 30357494
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357494 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357494 Supplier Toronto Research Chemicals Supplier No. TRCD2996085G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dextrin
CAS 9004-53-9
Molecular Formula (C14 H26 O10)n
Quantity 5 g
Synonym 102D, 102K, 1752S, 30AN, 37LAC19, 955SR, Amaizo 1706, Dextrid, Dextrin 10, Dextrin 101, Dextrin 102N, Dextrin 102S, Dextrin 103, Dextrin 1104, Dextrin 12, Dextrin 1719, Farinex CO 2, Farinex MJ, Fibersol 2, Fortodex, Fungal amylase starch, GLD, GOS-sugar, Globe 8051, Leiocom, Licopol, Ligament BAE 4, Ligament P 950A, Lycatab DSH, M 250, M 365, M 550, M 600, Sandec 100, Sandec 100T, Sandec 150, Sandec 250, Sandec 30, Sandec 300, Sandec 70, Tackidex 035, Tackidex 133, Tackidex C 172, Tackidex DF 165, Tackidex G 155
InChI Formula InChI=1S/C18H32O16/c19-1-4-7(22)8(23)12(27)17(31-4)34-15-6(3-21)32-18(13(28)10(15)25)33-14-5(2-20)30-16(29)11(26)9(14)24/h4-29H,1-3H2/t4-,5-,6-,7-,8+,9-,10-,11-,12-,13-,14-,15-,16+,17?,18?/m1/s1
SMILES C([C@@H]1[C@H]([C@@H]([C@H](C(O1)O[C@@H]2[C@H](OC([C@@H]([C@H]2O)O)O[C@@H]3[C@H](O[C@@H]([C@@H]([C@H]3O)O)O)CO)CO)O)O)O)O
Molecular Weight (g/mol) 162.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.