missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran, Ultrapure
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 1 kg |
| Formula Weight | 60,000- 90,000 |
| Packaging | Plastic bottle |
| Chemical Name or Material | Dextran |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction