missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate Sodium Salt (White to Off-white Powder), Fisher BioReagents

Product Code. 10295153
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295153 100 g Glass Bottle 100g
10400274 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295153 Supplier Fisher BioReagents Supplier No. BP1585100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Dextran Sulfate accelerates the rate of hybridization of DNA or RNA bound to membranes and may be used in Southern and Northern blotting procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 500,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name ethyl 4-aminobenzoate
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Color White
Physical Form Solid
Merck Index 15, 2953
Quantity 100 g
Identification Pass Test
Moisture 7.5% max.
Packaging Glass Bottle
Chemical Name or Material Dextran Sulfate Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.