missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran Sulfate, Sodium Salt, Ultrapure, Thermo Scientific™
£48.70 - £100.01
Chemical Identifiers
CAS | 9011-18-1 |
---|---|
Molecular Formula | (C6H7O5)m(C6H7O5)n |
Molecular Weight (g/mol) | 40,000 |
MDL Number | MFCD00081551 |
InChI Key | BLFLLBZGZJTVJG-UHFFFAOYSA-N |
Synonym | benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin |
PubChem CID | 2337 |
ChEBI | CHEBI:116735 |
IUPAC Name | ethyl 4-aminobenzoate |
SMILES | *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
9011-18-1 | |
40,000 | |
BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
2337 | |
ethyl 4-aminobenzoate |
(C6H7O5)m(C6H7O5)n | |
MFCD00081551 | |
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
CHEBI:116735 | |
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
Specifications
9011-18-1 | |
(C6H7O5)m(C6H7O5)n | |
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] | |
40,000 | |
CHEBI:116735 | |
Dextran sulfate sodium salt |
Powder | |
MFCD00081551 | |
BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
ethyl 4-aminobenzoate | |
2337 | |
500,000 MW |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures