missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate Sodium Salt Reagent MP Biomedicals

Product Code. 11427870 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427870 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11427870 Supplier MP Biomedicals Supplier No. 0210151601
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gold standard reagent for the induction of ulcerative colitis models in research animals.

Specifications

CAS 9011-18-1
Quantity 1 g
Molecular Formula (C6H7O5)m(C6H7O5)n
MDL Number MFCD00081551
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
IUPAC Name Dextran sulfate sodium salt
Molecular Weight (g/mol) 40,000
PubChem CID 2337
ChEBI CHEBI:116735
Formula Weight 8,000 (Average)
Grade Reagent
Chemical Name or Material Dextran Sulfate Sodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.