missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000

Product Code. 13505010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13505010 25 g 25g
13595000 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13505010 Supplier Thermo Scientific Acros Supplier No. 441490250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dextran sulfate sodium salt, C9H11NO2, CAS Number-9011-18-1, americaine, anaesthesin, anesthesin, benzocaine, ethoform, ethyl aminobenzoate, ethyl p-aminobenzoate, norcaine, orthesin, parathesin, 25g, CHEBI:116735, BLFLLBZGZJTVJG-UHFFFAOYSA-N, ethyl 4-aminobenzoate, MFCD00081551, 165.192 | CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Solubility Information Solubility in water: 100g/L (25°C).
Quantity 25 g
IUPAC Name ethyl 4-aminobenzoate
Packaging Plastic bottle
Chemical Name or Material Dextran sulfate sodium salt, approx. M.W. 6, 500 - 10, 000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.