missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate Sodium Salt (6,000-8,000 M.Wt), MP Biomedicals

Product Code. 15586226 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15586226 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15586226 Supplier MP Biomedicals Supplier No. 0210151890
  • ONLINE EXCLUSIVE PRICE
    £1835.00
    £1716.75 / 500g
    Save £118.25
    6% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Versatile polyanionic derivative of dextran used for research in immunology, antiviral, and antilipemic applications.

  • Dextran Sulfate Sodium (DSS) which has proven to have versatile value in Biomedical/clinical research
  • DSS can be used as an anticoagulant, antiviral as well as antilipemic agent

Specifications

Color White
CAS 9011-18-1
Physical Form Powder
Quantity 500 g
Molecular Formula (C6H7O5)m(C6H7O5)n
MDL Number MFCD00081551
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
Solubility Information Soluble in water (100mg/mL - clear to slightly hazy yellow solution.
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
IUPAC Name Dextran sulfate sodium salt
Molecular Weight (g/mol) 40,000
PubChem CID 2337
ChEBI CHEBI:116735
Formula Weight 6,000-8,000
Grade Not Available
Chemical Name or Material DEXTRAN SULFATE SODIUM SALT (6,000-8,000 M.Wt)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.