missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran sulfate sodium salt (400,000-600,000 M.Wt), MP Biomedicals™
Shop All MP Biomedicals ProductsSpecifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 1 g |
| IUPAC Name | Dextran sulfate sodium salt |
| Formula Weight | 400,000 to 600,000 |
| Chemical Name or Material | Dextran Sulfate Sodium Salt |
Safety and Handling
Product Identifier
- Dextran Sulfate Sodium Salt
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Dextran Sulfate Sodium Salt
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?