missing translation for 'onlineSavingsMsg'
Learn More

Dextran, MW ca 40,000, Thermo Scientific™

Dextran, CAS # 9004-54-0, is a glucose polymer. It is mainly used to increase electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity).

£28.65 - £503.65

Chemical Identifiers

CAS 9004-54-0
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYSA-N
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
PubChem CID 4125253
IUPAC Name 2,3,4,5-tetrahydroxy-6-[3,4,5-trihydroxy-6-[[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxyhexanal
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC(C(C(C(C=O)O)O)O)O)O)O)O)O)O)O)O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15445869
SDS View Documents
Thermo Scientific Alfa Aesar
J63690.09
10g
ONLINE EXCLUSIVE PRICE
£31.50
£28.65 / 10g
Save £2.85
9% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15455869
SDS View Documents
Thermo Scientific Alfa Aesar
J63690.18
50g
ONLINE EXCLUSIVE PRICE
£96.00
£86.65 / 50g
Save £9.35
9% Off
View alternative products
Save Up to 
»
In Stock
Edge
Add to basket
 
 Restricted Item
15465869
SDS View Documents
Thermo Scientific Alfa Aesar
J63690.22
100g
£143.00
100g
View alternative products
Save Up to 
»
In Stock
Edge
Add to basket
 
 Restricted Item
15475869
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
J63690.36
500g
ONLINE EXCLUSIVE PRICE
£559.00
£503.65 / 500g
Save £55.35
9% Off
View alternative products
Save Up to 
»
Estimated Shipment: 08-10-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

  • Dextran is a glucose polymer biosynthesized by the non-pathogenic Leuconostoc mesenteroides

Application

  • Dextran is a complex branched glucan that can be used to enhance electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity)
  • This compound promotes the activation of plasminogen by inhibiting α-2 antiplasmin
  • Dextran can be used to reduce thrombosis in animal models
  • It is used in osmotic stress techniques where osmotic pressure is applied to biological molecules
  • Dextran can be used as an immobilization agent in biosensors
Specifications

Chemical Identifiers

9004-54-0
504.438
FZWBNHMXJMCXLU-UHFFFAOYSA-N
4125253
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC(C(C(C(C=O)O)O)O)O)O)O)O)O)O)O)O
C18H32O16
MFCD00130935
dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
2,3,4,5-tetrahydroxy-6-[3,4,5-trihydroxy-6-[[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxyhexanal

Specifications

9004-54-0
MFCD00130935
dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
FZWBNHMXJMCXLU-UHFFFAOYSA-N
2,3,4,5-tetrahydroxy-6-[3,4,5-trihydroxy-6-[[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxyhexanal
4125253
Dextran
C18H32O16
14,2948
Soluble in water,dimethyl sulfoxide,ethylene glycol and glycerol.
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC(C(C(C(C=O)O)O)O)O)O)O)O)O)O)O)O
504.438
≈40 kd
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 232-677-5

missing translation for 'rtecsNumber' : HH9246000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.