missing translation for 'onlineSavingsMsg'
Learn More

Dextran, MP Biomedicals™

Product Code. 11490676 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490676 10 g 10g
11410686 100 g 100g
11420686 500 g 500g
11430686 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11490676 Supplier MP Biomedicals Supplier No. 0218014010
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Improves performance of immobilized proteins acting on macromolecules. Dextran, MP Biomedicals is a branched polysaccharide made up of linear α (1→6) linked glucose units and α (1→3) link initiated branches. It ranges in size from 10

Depending upon its molecular weight it has different uses in different fields.

  • May be used as long hydrophilic spacer arms to improve the freedom of movement of conjugated/bound proteins.
  • May be derivatized for use in biosensor systems
  • Other applications include: platelet aggregant, plasma volume extender, osmotic pressure regulator, stabilizer, organ separation media, matrix component, copolymer, microcarrier, binding agent, viscosity modifier, antithrombotic, lubricant and physical structure component.

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
PubChem CID 4125253
IUPAC Name 2,3,4,5-tetrahydroxy-6-[3,4,5-trihydroxy-6-[[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxyhexanal
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Very soluble in water (50mg/mL), slightly soluble in alcohol.
Formula Weight 60,000-90,000
Grade Clinical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.