missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexras1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £359.00
Specifications
| Antigen | Dexras1 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668171
|
Novus Biologicals
NBP2-92235-0.02ml |
0.02 mL |
£153.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686631
|
Novus Biologicals
NBP2-92235-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dexras1 Polyclonal antibody specifically detects Dexras1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| Dexras1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 51655 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Activator of G-protein signaling 1, AGS1ras-related protein, DEXRAS1dexamethasone-induced Ras-related protein 1, MGC:26290, RAS, dexamethasone-induced 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 132-281 of human RASD1 (NP_057168.1). TKENVDVPLVICGNKGDRDFYREVDQREIEQLVGDDPQRCAYFEISAKKNSSLDQMFRALFAMAKLPSEMSPDLHRKVSVQYCDVLHKKALRNKKLLRAGSGGGGGDPGDAFGIVAPFARRPSVHSDLMYIREKASAGSQAKDKERCVIS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title