missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ DexPure Vinyl Gloves
Description
- Translucent vinyl glove.
- Smooth finish for great dexterity.
- Certified for food contact and treated to prevent bacteria.
- Suitable for the following:
- Product inspection.
- Precision assembly.
- Laboratory work.
- Handling of paints, inks and dyes.
- Single use.
- AQL:1.5.
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 374-2003 Chemical hazards, EN 455-1&2 Medical Standards, European directive n°1934/2004 : Food contact, EC Category PPE 3, EC Certificate Number 0075/797/162/11/08/0465, EC Attestation Number 0075/797/162/11/08/0465 |
| Classification | Dexterity: 5 |
| Description | AQL:1.5 |
| Coating Material | Vinyl |
| Color | Translucent |
| Disposable | Single-use |
| Material | Vinyl |
| Powdered | Powdered |
| Product Type | Gloves |
| Size | Large |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?