missing translation for 'onlineSavingsMsg'
Learn More

Dexpramipexole dihydrochloride, MedChemExpress

Product Code. p-200100173
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16457298

Brand: Medchem Express HY17355A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dexpramipexole dihydrochloride ((R)-Pramipexole dihydrochloride) is a neuroprotective agent and weak non-ergoline dopamine agonist.

Dexpramipexole dihydrochloride ((R)-Pramipexole dihydrochloride) is a neuroprotective agent and weak non-ergoline dopamine agonist.

Specifications

Chemical Name or Material Dexpramipexole dihydrochloride
Physical Form Solid
CAS 104632-27-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H19Cl2N3S
For Use With (Application) Neuroscience-Neurodegeneration
Synonym (R)-Pramipexole dihydrochlorideR-(+)-Pramipexole dihydrochlorideKNS-760704 dihydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (351.80 mM) ∣H2O : 100 mg/mL (351.80 mM; Need ultrasonic)
SMILES NC(S1)=NC2=C1C[C@H](NCCC)CC2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 284.25
Formula Weight 284.25
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.