missing translation for 'onlineSavingsMsg'
Learn More

Dexpanthenol, Mikromol™

Product Code. 17910035
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17910035 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17910035 Supplier Mikromol Supplier No. MM1352.000250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dexpanthenol
TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material D-Panthenol
CAS 81-13-0
Analyte or Component Names D-Panthenol
Shipping Condition Room Temperature
Molecular Formula C9 H19 N O4
Certifications/Compliance ISO 17034
Synonym (2R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide, D-(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, (+)-Panthenol, Alcopan 250, Bepanthen, Bepanthene, Bepantol, Cornergel, Cozyme, D(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, D(+)-Panthenol, D(+)-Pantothenyl alcohol, D(+)-alpha,gamma-Dihydroxy-N-(3-hydroxypropyl)-beta,beta-dimethylbutyramide, D-P-A Injection, D-Panthenol, D-Pantothenyl alcohol, Dexpanthenol, Dolobene, Ilopan, Intrapan, Motilyn, NSC 302962, Panadon, Pantenyl, Panthenol, Panthenol 50W, (+)-Panthenol, Panthoderm, Pantol, Pantothenol, Pantothenyl alcohol, N-Pantoylpropanolamine, Provitamin B, Provitamin B5, Synapan, Thenalton, Urupan, Zentinic, d-Panthenol, d-Panthenol 50, d-Pantothenol, d-Pantothenyl alcohol
InChI Formula InChI=1S/C9H19NO4/c1-9(2,6-12)7(13)8(14)10-4-3-5-11/h7,11-13H,3-6H2,1-2H3,(H,10,14)/t7-/m0/s1
SMILES CC(C)(CO)[C@@H](O)C(=O)NCCCO
IUPAC Name (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
Molecular Weight (g/mol) 205.25
Quantity 250 mg
Formula Weight 205.1314 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.