missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexmedetomidine hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Dexmedetomidine hydrochloride ((+)-Medetomidine hydrochloride) is a potent, selective and orally active agonist of α2-adrenoceptor, with a Ki of 1.08 nM. Dexmedetomidine hydrochloride shows 1620-fold selectivity against α1-adrenoceptor. Dexmedetomidine hydrochloride exhibits anxiolysis, sedation, and modest analgesia effects.
Specifications
Specifications
| Chemical Name or Material | Dexmedetomidine hydrochloride |
| Physical Form | Solid |
| CAS | 145108-58-3 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C13H17ClN2 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | (+)-Medetomidine hydrochloride(S)-Medetomidine hydrochloride |
| Solubility Information | DMSO : 250 mg/mL (1056.01 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (211.20 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction