missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEXI Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116601
This item is not returnable.
View return policy
Description
Positive Samples: A-549, BxPC-3, HT-29 Immunogen sequence: MLGARVAAHL DALGPLVPYV PPPLLPSMFY VGLFFVNVLI LYYAFLMEYI VLNVGLVFLP EDMDQALVDL GVLSDPGSGL YDADSELDVF DAYLE.
DEXI (dexamethasone-induced protein), also known as MYLE, is a 95 amino acid protein belonging to the DEXI family.
Specifications
| DEXI | |
| Polyclonal | |
| Unconjugated | |
| Dexi | |
| 1810029J14Rik; AI836170; AW413143; D16Bwg0586e; Dexamethasone-induced protein; dexamethasone-induced transcript; Dexi; Dexi homolog; Myle; Protein MYLE; RGD1564938 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 28955 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.21 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O95424 | |
| Dexi | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-95 of human DEXI (NP_0547342). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction