missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone, TRC

Product Code. 18052400
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18052400 5 g 5g
18032000 25 g 25g
18020190 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18052400 Supplier Toronto Research Chemicals Supplier No. TRCD2988005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-02-2
Molecular Formula C22 H29 F O5
Molecular Weight (g/mol) 392.46
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl- (6CI,8CI), (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 1-Dehydro-16α-methyl-9α-fluorohydrocortisone, 16α-Methyl-9α-fluoro-1,4-pregnadiene-11β,17α,21-triol-3,20-dione, 16α-Methyl-9α-fluoro-11β,17α,21-trihydroxypregna-1,4-diene-3,20-dione, 16α-Methyl-9α-fluoro-δ1-hydrocortisone, 16α-Methyl-9α-fluoroprednisolone, 9-Fluoro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-11β,17α,21-trihydroxy-16α-methyl-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methyl-1,4-pregnadiene-11β,17α,21-triol-3,20-dione, 9α-Fluoro-16α-methyl-11β,17,21-trihydroxypregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Adexone, Aeroseb-Dex, Aphtasolon, Aphthasolone, Azium, Calonat, Corsone, Cortisumman, Decacort, Decaderm, Decadron, Decadron A, Decalix, Decasone, Dekacort, Dekort, Delipos, Deltafluorene, Dergramin, Deronil, Desadrene, Desameton, Deseronil, Dexa-Cortidelt, Dexa-Mamallet, Dexa-Scheroson, Dexa-sine, Dexacort, Dexacortal, Dexacortin, Dexadeltone, Dexafarma, Dexalona, Dexaltin, Dexameth, Dexamethasone, Dexamethasone alcohol, Dexamonozon, Dexapolcort, Dexapos, Dexaprol, Dexaratiopharm, Dexason, Dexasone, Dexaven, Dexinoral, Dexium, Dexmethsone, Dexone, Dexonium, Dextelan, Dinormon, Diodex, Etacortilen, Fluormethylprednisolone, Fluormone, Fluorocort, Gammacorten, Gentalipos, HL-Dex, Hexadecadrol, Hexadrol, Isopto-Dex, Lokalison F, Loverine, Luxazone, MK 125, Maxidex, Millicorten, Minidex, NSC 34521, Oradexon, Ozurdex, Pet-Derm III, Posurdex, Prednisolon F, Prednisolone F, Superprednol, Surodex, Visumetazone, Betamethasone Imp. A (EP), Dexamethasone Acetate Imp. A (EP), Dexamethasone Sodium Phosphate Imp. A (EP), Dexamethasone, 9-Fluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione, Betamethasone Impurity A, Dexamethasone Acetate Impurity A, Dexamethasone Isonicotinate Impurity A, Dexamethasone Sodium Phosphate Impurity A
IUPAC Name (8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1
Formula Weight 392.1999
Percent Purity >95%
Chemical Name or Material Dexamethasone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.