missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone Sodium Phosphate, Mikromol™

Product Code. 17939709
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17939709

Brand: Mikromol MM0210.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dexamethasone Sodium Phosphate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dexamethasone disodium phosphate
CAS 2392-39-4
Analyte or Component Names Dexamethasone disodium phosphate
Shipping Condition Room Temperature
Molecular Formula C22 H28 F O8 P . 2 Na
Certifications/Compliance ISO 17034
Synonym Dexamethasone Sodium Phosphate, Ak-Dex, Baldex, Colvasone, Corson, Corson (pharmaceutical), Dalalone, Decadron phosphate, Decdan, Dexabene, Dexacort, Dexadreson, Dexagel, Dexagro, Dexamethasone 21-(disodium phosphate), Dexamethasone 21-phosphate disodium salt, Dexamethasone disodium phosphate, Dexamethasone phosphate disodium salt, Dexamethasone phosphate sodium salt, Dexamethasone sodium phosphate, Dexamethazone sodium phosphate, Dezone, Disodium dexamethasone 21-monophosphate, Disodium dexamethasone 21-phosphate, Disodium dexamethasone phosphate, Maxidex Ointment, Megacort, Onadron, Orgadrone, Sodium dexamethasone phosphate, Soldesam, Solu-Deca, Solu-Decadron, Spersadex, Turbinaire, (11beta,16alpha)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione sodium salt (1:2), (11beta,16alpha)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione disodium salt, 9-Fluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione 21-(dihydrogen phosphate) disodium salt, 9-Fluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione 21-phosphate disodium salt
InChI Formula InChI=1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15+,16+,17+,19+,20+,21+,22+;;/m1../s1
SMILES [Na+].[Na+].C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP(=O)([O-])[O-]
IUPAC Name disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate
Molecular Weight (g/mol) 516.4
Quantity 250 mg
Formula Weight 516.1301 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.